missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Danshensu, MedChemExpress
Description
Danshensu, an active ingredient of Salvia miltiorrhiza, shows wide cardiovascular benefit by activating Nrf2 signaling pathway.
Specifications
Specifications
| Chemical Name or Material | Danshensu |
| CAS | 76822-21-4 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C9H10O5 |
| Quantity | 10 mg |
| Synonym | Dan shen suan A Salvianic acid A |
| Solubility Information | H2O : 5 mg/mL (25.23 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | OC1=C(O)C=CC(C[C@@H](O)C(O)=O)=C1 |
| Molecular Weight (g/mol) | 198.17 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?