Learn More
Dalbavancin hydrochloride, MedChemExpress
Product Code. 16467408
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Product Code. 16467408
Brand: Medchem Express HY17586
MedChemExpress Dalbavancin hydrochloride (MDL-63397 hydrochloride) is a semisynthetic lipoglycopeptide antibiotic with potent bactericidal activity against Gram-positive bacteria. Dalbavancin hydrochloride inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 μg/mL and 0.25 μg/mL, respectively.
Dalbavancin hydrochloride (MDL-63397 hydrochloride) is a semisynthetic lipoglycopeptide antibiotic with potent bactericidal activity against Gram-positive bacteria. Dalbavancin hydrochloride inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 μg/mL and 0.25 μg/mL, respectively.
Specifications
Chemical Name or Material Dalbavancin hydrochloride
Physical Form Solid
CAS 2227366-51-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C88H101Cl3N10O28
For Use With (Application) COVID-19-immunoregulation
Synonym MDL-63397 hydrochlorideBI-397 hydrochloride
Solubility Information DMSO : 250 mg/mL (134.91 mM; Need ultrasonic) ∣H2O : 50 mg/mL (26.98 mM; Need ultrasonic)
SMILES O[C@@H]([C@@H](O)[C@@H]1O)[C@H](O[C@@H]1CO)OC2=C(C3=CC([C@@](C4=O)([H])NC([C@@](NC([C@](NC([C@@](NC5=O)([H])C6)=O)([H])C(C=C7OC(C=C8[C@H]5NC)=C(C=C8)O)=C(C(O)=C7)Cl)=O)([H])C9=CC%10=C(O[C@H](O[C@H](C(O)=O)[C@@H](O)[C@@H]%11O)[C@@H]%11NC(CCCCCCCCC(C)C)=O)C(OC%12=CC=C6C=C%12)=C9)=O)=CC=C3O)C([C@@](NC([C@](N4)([H])[C@@H](C%13=CC=C(O%10)C(Cl)=C%13)O)=O)([H])C(NCCCN(C)C)=O)=CC(O)=C2.[H]Cl
Molecular Weight (g/mol) 1853.15
Formula Weight 1853.15
Percent Purity 95.92%
Grade Research