Learn More
Memmert™ DAkkS Factory Calibration for UF750
Product Code. 17549555 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17549555
Brand: Memmert™ UF750E39696
DAkkS factory calibration for a freely selectable temperature for Memmert Ovens.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) UF750 Oven
Product Type DAkkS Factory Calibration