Learn More
Daclatasvir RSSR Isomer, TRC
Specifications
Specifications
| Chemical Name or Material | Daclatasvir RSSR Isomer |
| CAS | 1009107-27-0 |
| Molecular Formula | C40 H50 N8 O6 |
| Quantity | 100 mg |
| Synonym | N,N'-[[1,1'-Biphenyl]-4,4'-diylbis[1H-imidazole-5,2-diyl-(2S)-2,1-pyrrolidinediyl[(1R)-1-(1-methylethyl)-2-oxo-2,1-ethanediyl]]]biscarbamic Acid C,C'-Dimethyl Ester |
| InChI Formula | InChI=1S/C40H50N8O6/c1-23(2)33(45-39(51)53-5)37(49)47-19-7-9-31(47)35-41-21-29(43-35)27-15-11-25(12-16-27)26-13-17-28(18-14-26)30-22-42-36(44-30)32-10-8-20-48(32)38(50)34(24(3)4)46-40(52)54-6/h11-18,21-24,31-34H,7-10,19-20H2,1-6H3,(H,41,43)(H,42,44)(H,45,51)(H,46,52)/t31-,32-,33?,34?/m1/s1 |
| SMILES | COC(=O)NC(C(C)C)C(=O)N1CCC[C@@H]1c2ncc([nH]2)c3ccc(cc3)c4ccc(cc4)c5cnc([nH]5)[C@H]6CCCN6C(=O)C(NC(=O)OC)C(C)C |
| IUPAC Name | methyl N-[1-[(2R)-2-[5-[4-[4-[2-[(2R)-1-[2-(methoxycarbonylamino)-3-methylbutanoyl]pyrrolidin-2-yl]-1H-imidazol-5-yl]phenyl]phenyl]-1H-imidazol-2-yl]pyrrolidin-1-yl]-3-methyl-1-oxobutan-2-yl]carbamate |
| Molecular Weight (g/mol) | 738.875 |
| Formula Weight | 738.385 |