Learn More
Dabigatran-d4 hydrochloride, MedChemExpress
Product Code. 16476197
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16476197
Brand: Medchem Express HY10163AS
MedChemExpress Dabigatran (BIBR-953) D4 hydrochloride is deuterium labeled Dabigatran, which is a reversible and selective, direct thrombin inhibitor (DTI) with a Ki value of 4.5 nM.
Dabigatran (BIBR-953) D4 hydrochloride is deuterium labeled Dabigatran, which is a reversible and selective, direct thrombin inhibitor (DTI) with a Ki value of 4.5 nM.
Specifications
Chemical Name or Material Dabigatran-d4 hydrochloride
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C25H22D4ClN7O3
Color Colorless
Synonym BIBR-953-d4 hydrochloride
SMILES N=C(N)C(C([2H])=C1[2H])=C([2H])C([2H])=C1NCC2=NC(C=C3C(N(CCC(O)=O)C4=NC=CC=C4)=O)=C(C=C3)N2C.Cl
Industry Type Neuroscience-Neuromodulation
Molecular Weight (g/mol) 512.0
Quantity 5 mg
Formula Weight 512.0
Percent Purity 97.05%
Grade Research