missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran Acyl-Beta-D-Glucuronide Trifluoroacetic Acid Salt (80%), TRC

Product Code. 30009617
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009617 10 mg 10mg
30012374 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009617 Supplier Toronto Research Chemicals Supplier No. TRCD10008510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C31H33N7O9 • x(C2HF3O2)
Molecular Weight (g/mol) 647.6411402
Synonym N-[[2-[[[4-(Aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine β-D-Glucopyranuronosyl Ester Trifluoroacetic Acid Salt;
SMILES O=C(C1=[C]C=C2C(N=C(CNC3=CC=C(C(N)=N)C=C3)N2C)=C1)N(CCC(O[C@@H]4O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]4O)=O)C5=NC=CC=C5

Specifications

Analyte or Component Names Dabigatran Acyl-beta-D-Glucuronide Trifluoroacetic Acid Salt
Shipping Condition Dry Pack
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Chemical Name or Material Dabigatran Acyl-beta-D-Glucuronide Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.