Learn More
3M™ Secure Click™ Particulate Filter P1 R, D7915
Description
- New dual flow cartridges and filters allow for easier breathing
- Protects against particulates
- Quick, simple connection. Simply align the connections and push until you hear a click
- Use with 3M™ Secure Click™ Reusable Respirators 800 Series
- Swept-back design allows an enhanced field of view and comfort
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Connection Type | Secure Click™ |
| Filter Type | P1R |
| Product Type | Replacement Filter |
| For Use With (Equipment) | 3M™ Secure Click™ Respirators 800 Series, Secure Click™ Filter Cartridges D8000 Series, and Filter Retainer D701 |
| Model | D7915 |