missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ D4-GDI Monoclonal Antibody (10D774)

Description
Suggested positive control: antigen standard for ARHGDIB (transient overexpression lysate), Daudi or Jurkat.
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications
Specifications
| Antigen | D4-GDI |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 10D774 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% gelatin and 0.02% sodium azide |
| Gene | ARHGDIB |
| Gene Accession No. | P52566 |
| Gene Alias | ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2 |
| Gene Symbols | ARHGDIB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?