missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-abiraterone, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
D4-abiraterone is a major metabolite of abiraterone. D4-abiraterone is an inhibitor of CYP17A1, 3b-hydroxysteroid dehydrogenase (3βHSD) and steroid-5a-reductase (SRD5A) and also an antagonist of androgen receptor.
Specifications
Specifications
| Chemical Name or Material | D4-abiraterone |
| Physical Form | Solid |
| CAS | 154229-21-7 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H29NO |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | δ4-Abiraterone CB-7627 Abiraterone D4A metabolite |
| Solubility Information | DMSO : 50 mg/mL (143.89 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction