missing translation for 'onlineSavingsMsg'
Learn More

D4-abiraterone, MedChemExpress

Product Code. 16454318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
16454318 10 mg 10mg
16463036 25 mg 25mg
16447037 5 mg 5mg
3 options
This item is not returnable. View return policy

Product Code. 16454318

Brand: Medchem Express HY109619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D4-abiraterone is a major metabolite of abiraterone. D4-abiraterone is an inhibitor of CYP17A1, 3b-hydroxysteroid dehydrogenase (3βHSD) and steroid-5a-reductase (SRD5A) and also an antagonist of androgen receptor.

D4-abiraterone is a major metabolite of abiraterone. D4-abiraterone is an inhibitor of CYP17A1, 3b-hydroxysteroid dehydrogenase (3βHSD) and steroid-5a-reductase (SRD5A) and also an antagonist of androgen receptor.

Specifications

Chemical Name or Material D4-abiraterone
Physical Form Solid
CAS 154229-21-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H29NO
For Use With (Application) Cancer-programmed cell death
Synonym δ4-Abiraterone CB-7627 Abiraterone D4A metabolite
Solubility Information DMSO : 50 mg/mL (143.89 mM; Need ultrasonic)
SMILES C[C@@]12C(C3=CC=CN=C3)=CC[C@@]1([H])[C@]4([H])CCC5=CC(CC[C@]5(C)[C@@]4([H])CC2)=O
Molecular Weight (g/mol) 347.49
Formula Weight 347.49
Percent Purity 99.27%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.