missing translation for 'onlineSavingsMsg'
Learn More

D3-βArr, MedChemExpress

Product Code. p-200098275
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495198

Brand: Medchem Express HY124867

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D3-βArr is a positive allosteric modulator for thyrotropin receptor (TSHR), which initiates translocation of β-Arr 1 by direct TSHR activation and potentiates TSH-mediated preosteoblast differentiation in vitro.

D3-βArr is a positive allosteric modulator for thyrotropin receptor (TSHR), which initiates translocation of β-Arr 1 by direct TSHR activation and potentiates TSH-mediated preosteoblast differentiation in vitro.

Specifications

Chemical Name or Material D3-βArr
Physical Form Solid
CAS 662164-09-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H23N5
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 125 mg/mL (374.89 mM)
SMILES CC(NC1=C2C=CC=CC2=NC(N3CCNCC3)=N1)C4=CC=CC=C4
Molecular Weight (g/mol) 333.43
Formula Weight 333.43
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.