missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D3-βArr, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
D3-βArr is a positive allosteric modulator for thyrotropin receptor (TSHR), which initiates translocation of β-Arr 1 by direct TSHR activation and potentiates TSH-mediated preosteoblast differentiation in vitro.
Specifications
Specifications
| Chemical Name or Material | D3-βArr |
| Physical Form | Solid |
| CAS | 662164-09-2 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23N5 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : ≥ 125 mg/mL (374.89 mM) |
| SMILES | CC(NC1=C2C=CC=CC2=NC(N3CCNCC3)=N1)C4=CC=CC=C4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction