missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D12-050 Standard: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 50 μg/g
Specifications
Specifications
| Chemical Name or Material | Aluminium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Nickel∣Silicon∣Silver∣Sodium∣Tin∣Titanium |
| CAS | 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4 |
| Analyte or Component Names | Aluminium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Nickel∣Silicon∣Silver∣Sodium∣Tin∣Titanium |
| Shipping Condition | Room Temperature |
| Element | Multi-element |
| Industry Type | Petrochemical Testing |
| Quantity | 200 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 50 μg/g∣Chromium 50 μg/g∣Copper 50 μg/g∣Iron 50 μg/g∣Lead 50 μg/g∣Magnesium 50 μg/g∣Nickel 50 μg/g∣Silicon 50 μg/g∣Silver 50 μg/g∣Sodium 50 μg/g∣Tin 50 μg/g∣Titanium 50 μg/g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?