missing translation for 'onlineSavingsMsg'
Learn More

D12-005 Standard: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 5 μg/g

Product Code. 30325458
Change view
Click to view available options
Quantity:
200 g
Unit Size:
200g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325458 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325458 Supplier VHG Labs Supplier No. VHGD125200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Nickel∣Silicon∣Silver∣Sodium∣Tin∣Titanium
CAS 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4
Analyte or Component Names Aluminium∣Chromium∣Copper∣Iron∣Lead∣Magnesium∣Nickel∣Silicon∣Silver∣Sodium∣Tin∣Titanium
Shipping Condition Room Temperature
Element Multi-element
Industry Type Petrochemical Testing
Quantity 200 g
Concentration or Composition (by Analyte or Components) Silicon 5.01 μg/g∣Aluminium 5.03 μg/g∣Chromium 5.03 μg/g∣Copper 5.03 μg/g∣Iron 5.03 μg/g∣Lead 5.03 μg/g∣Magnesium 5.03 μg/g∣Nickel 5.03 μg/g∣Silver 5.03 μg/g∣Sodium 5.03 μg/g∣Tin 5.03 μg/g∣Titanium 5.03 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.