missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford D-(+)-Raffinose Pentahydrate
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Unit Size:
100g
1kg
25g
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Sulfated Ash: <0.1%
Heavy Metals: <0.001%
Specifications
Specifications
| CAS | 17629-30-0, 512-69-6 |
| Molecular Formula | C18H32O16-5H2O |
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction