Learn More
STEMCELL Technologies D-PBS (Without Ca++ and Mg++)
Product Code. 15252745
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15252745 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15252745 Supplier STEMCELL Technologies Supplier No. 37350
Dulbecco’s phosphate-buffered saline without calcium and magnesium
  • Dulbecco's phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating, or transporting solution for cell or tissue culture
  • It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions
Specifications
Quantity 500 mL