Learn More
STEMCELL Technologies D-PBS (Without Ca++ and Mg++)
Description
- Dulbecco's phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating, or transporting solution for cell or tissue culture
- It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions
Specifications
Specifications
| Quantity | 500 mL |