Learn More
D(+)-Fenchone, 97%
Product Code. 10180032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10180032 100 g Glass bottle 100g
10018771 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10180032 Supplier Thermo Scientific Acros Supplier No. 366291000
Request Bulk or Custom Format
CAS: 4695-62-9 | C10H16O | 152.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4695-62-9
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00070689
InChI Key LHXDLQBQYFFVNW-XCBNKYQSSA-N
Synonym +-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
PubChem CID 1201521
ChEBI CHEBI:165
IUPAC Name (1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C
Specifications
Melting Point 5°C to 7°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 63°C to 65°C (13.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.464
Quantity 100 g
Beilstein 07, IV, 212
Merck Index 15, 4000
Specific Gravity 0.95
Specific Rotation Condition +59.00 (20.00°C neat)
Specific Rotation + 59
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in ethanol,ether,common organic solvents
Formula Weight 152.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material D(+)-Fenchone, 97%