missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Cycloserine

Product Code. p-200087243
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018207

Brand: Melford C810005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine.

D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine. It inhibits cell wall synthesis in certain bacteria, including Mycobacterium tuberculosis. While it was initially identified as an antibiotic, D-cycloserine has gained attention for its activity as a partial agonist at the N-methyl-D-aspartate (NMDA) receptor, a type of glutamate receptor in the brain. Researchers study the mechanisms of neurotransmission and synaptic plasticity due to its interactions with NMDA receptors.

Specifications

Chemical Name or Material D-Cycloserine
CAS 68-41-7
Quantity 5 g
Molecular Formula C3H6N2O2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.