missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS

Product Code. 11308369 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11308369

Brand: MP Biomedicals 02100071.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytosine-β-D-Arabinofuranoside Hydrochloride

Application Notes:

  • It is used as an antineoplastic and antiviral agent.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Selective inhibitor of DNA synthesis
TRUSTED_SUSTAINABILITY

Specifications

Product Type Nucleoside
Quantity 500 mg
Color White
Molecular Formula C9H14ClN3O5
CAS 69-74-9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.