missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 14/16 Polyclonal Antibody, Invitrogen™

Product Code. 15557501
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15557501 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557501 Supplier Invitrogen Supplier No. PA536061

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 52 kDa. Purity is >95% by SDS-PAGE.

Cytokeratin 14/16 Polyclonal antibody specifically detects Cytokeratin 14/16 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14/16
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Krt16
Gene Accession No. P02533, P08779, Q61781, Q6IFV1, Q9Z2K1
Gene Alias AI324768; AI626930; CK 14; CK 16; CK14; CK-14; CK16; CK-16; cytokeratin 14; cytokeratin 16; Cytokeratin14; cytokeratin-14; Cytokeratin16; Cytokeratin-16; EBS3; EBS4; epidermolysis bullosa simplex, Dowling-Meara, Koebner; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; K14; K16; K1CP; Ka14; Ka16; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 16; keratin 16, type I; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 14; Keratin, type I cytoskeletal 16; keratin-14; Keratin-16; Krt-1.14; Krt1-14; Krt1-16; Krt14; KRT16; KRT16A; NEPPK; NFJ; PC1; Type I keratin Ka14; type I keratin KA16
Gene Symbols KRT14, Krt16
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human Cytokeratin 14
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16664, 16666, 287701, 303530, 3861, 3868
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.