missing translation for 'onlineSavingsMsg'
Learn More

Cytohesin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16896394 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16896394 20 μL 20µL
16886394 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16896394 Supplier Proteintech Supplier No. 166871AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GRP1 (general receptor for phosphoinositides-1) contains a Pleckstrin homology (PH) domain as well as a Sec7 domain. The PH domain has high binding affinity for phosphatidylinositol 3,4,5-trisphosphate Ptdlns(3,4,5)P3), while the Sec7 homology domain is responsible for catalyzing guanine nucleotide exchange of ADP-ribosylation factor (ARF) proteins. GRP1 co-localizes with ARF6 and catalyzes GTP/GDP exchange on ARF6. It is known to interact with Ptdlns(3,4,5)P3 localized to the plasma membrane in vitro and may also be a Ptdlns(3,4,5)P3 receptor. Additionally, GRP1 may regulate protein sorting and membrane trafficking through interaction with the guanosine triphosphate ARF, and may control cell adhesion through interaction with integrins.

Specifications

Antigen Cytohesin 3
Applications Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYTH3
Gene Accession No. O08967, O43739, P97696
Gene Alias ARNO3, CYTH3, cytohesin 3, GRP1, Protein ARNO3, PSCD3
Gene Symbols Cyth3
Host Species Rabbit
Immunogen Cytohesin 3 Fusion Protein Ag10025
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116693, 19159, 9265
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.