missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyproterone acetate, MedChemExpress
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Description
Cyproterone acetate is an anti-androgen (IC50=7.1 nM) and progestogen synthetic steroid. Cyproterone acetate has affinity with progesteron and with glucocorticoidal receptors.
Specifications
Specifications
| Chemical Name or Material | Cyproterone acetate |
| Physical Form | Solid |
| CAS | 427-51-0 |
| Color | Off-White |
| Quantity | 250 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H27ClO4 |
| Solubility Information | DMSO : 33.33 mg/mL (82.72 mM; Need ultrasonic) ∣Ethanol : 20 mg/mL (49.64 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.25 mM; Need ultrasonic) |
| SMILES | C[C@@]12C(C(Cl)=C[C@]3([H])[C@]2([H])C[C@@]4(C)[C@@]3([H])CC[C@@]4(C(C)=O)OC(C)=O)=CC([C@H]5[C@@H]1C5)=O |
| Molecular Weight (g/mol) | 402.91 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction