Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Cyphenothrin |
| CAS | 39515-40-7 |
| CAS Max % | ≤ 100% |
| Flash Point | 130°C |
| Color | Yellow |
| Synonym | (RS)-a-cyano-3-phenoxybenzyl (1 RS,3 RS;1 RS,3 SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate |
| Molecular Weight (g/mol) | 375.46 g/mol |
| Quantity | 100 mg |