Learn More
CYP20A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16836424 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16836424 20 μL 20µL
16826424 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16836424 Supplier Proteintech Supplier No. 167021AP20UL
Rabbit Polyclonal Antibody
CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.
Specifications
Antigen CYP20A1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP20A1
Gene Accession No. Q6P7D4, Q6UW02, Q8BKE6
Gene Alias CYP M, CYP20A1, Cytochrome P450 20A1
Gene Symbols Cyp20a1
Host Species Rabbit
Immunogen CYP20A1 Fusion Protein Ag10099
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316435, 57404, 77951
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG