Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polypropylene
Description
- For laboratory use
- For collection and evaluation of samples
- Caps are not autoclavable
Specifications
Specifications
| Closure Color | Red |
| Color | Clear |
| Capacity (Metric) | 15 mL |
| Length (Metric) | 100 mm |
| Material | Polypropylene |
| Shape | Cylindrical |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Graduated | No |
| Show More |