Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Unit Size:
Each
Pack of 2
Description
- With scale (approx. volume/mL)
- Without pressure equalizing tube
- With screwthread retaining device
- Cylindrical shape with NS socket and cone
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Graduated | Yes |
| Includes | w/o Pressure Equalizing Tube |
| Joint Number | 29/32 NS |
| Product Type | Dropping Funnel |