Learn More
Thermo Scientific Chemicals Al-24 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20
Product Code. 15433525
Click to view available options
Capacity (Metric):
6 mL
20 mL
40 mL
60 mL
110 mL
225 mL
600 mL
Diameter (Metric) Outer:
16mm
26mm
30mm
36mm
50mm
55mm
73mm
Diameter (Metric) Inner:
30mm
40mm
50mm
60mm
75mm
100mm
150mm
Height (Metric):
20mm
30mm
35mm
40mm
65mm
75mm
85mm
missing translation for 'unitSize'
Each
Product Code. 15433525
missing translation for 'mfr': Thermo Scientific Chemicals 032966.KF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 100mm
Material Al-24
Quantity 1 Piece
Capacity (Metric) 225 mL
Diameter (Metric) Outer 55mm
Height (Metric) 65mm
Product Type Cylindrical Al-24 Crucibles
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.