Learn More
Saint-Gobain Cylinder measuring spouted PFA graduated 300mL Chemware
Brand:  Saint-Gobain D1069605
Additional Details : Weight : 0.01000kg
Product Code. 12388915
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Graduated Cylinder
300 mL