missing translation for 'onlineSavingsMsg'
Learn More

Cyhalodiamide, TRC

Product Code. 30134711
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134711 250 mg 250mg
17980595 100 mg 100mg
17973825 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134711 Supplier Toronto Research Chemicals Supplier No. TRCC937260250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262605-53-7
Molecular Formula C22 H17 Cl F7 N3 O2
Molecular Weight (g/mol) 523.83
IUPAC Name 3-chloro-2-N-(2-cyanopropan-2-yl)-1-N-[4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-2-methylphenyl]benzene-1,2-dicarboxamide
SMILES Cc1cc(ccc1NC(=O)c2cccc(Cl)c2C(=O)NC(C)(C)C#N)C(F)(C(F)(F)F)C(F)(F)F

Specifications

Quantity 250 mg
Synonym 3-Chloro-N2-(1-cyano-1-methylethyl)-N1-[2-methyl-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]phenyl]-1,2-benzenedicarboxamide, 3-Chloro-N2-(2-cyanopropan-2-yl)-N1-[4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-2-methylphenyl]phthalamide, Cyhalodiamide, ZJ 4042, ZJ4042
InChI Formula InChI=1S/C22H17ClF7N3O2/c1-11-9-12(20(24,21(25,26)27)22(28,29)30)7-8-15(11)32-17(34)13-5-4-6-14(23)16(13)18(35)33-19(2,3)10-31/h4-9H,1-3H3,(H,32,34)(H,33,35)
Formula Weight 523.0898
Percent Purity >95%
Chemical Name or Material Cyhalodiamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.