Learn More
Cyclohexanone oxime, 97%
Product Code. p-96655 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
500g
Product Code. 10112620
Brand: Thermo Scientific Acros 111201000
Request Bulk or Custom Format
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1
Specifications
Melting Point 88°C to 92°C
Color Beige-Brown to White
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Crystalline Powder, Crystals, or Chunks
Assay Percent Range 97%
Linear Formula C6H10(=NOH)
Beilstein 07, 10
Merck Index 10, 2720
Formula Weight 113.16
Percent Purity ≥96.0%
Chemical Name or Material Cyclohexanone oxime, 97%