missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanemethanol, TRC

Product Code. 17920095
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920095 5 g 5g
17998514 100 g 100g
17939754 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920095 Supplier Toronto Research Chemicals Supplier No. TRCC9891155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-49-2
Formula Weight 114.1045
InChI Formula InChI=1S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8H,1-6H2
IUPAC Name cyclohexylmethanol
Molecular Formula C7 H14 O
Molecular Weight (g/mol) 114.19
Percent Purity >95%
SMILES OCC1CCCCC1
Synonym Cyclohexanemethanol, (Hydroxymethyl)cyclohexane, Cyclohexanecarbinol, Cyclohexylcarbinol, Cyclohexylmethanol, Cyclohexylmethyl alcohol, Hexahydrobenzyl alcohol, NSC 5288
Chemical Name or Material Cyclohexylmethanol
Grade GC
Show More Show Less

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.