Learn More
Cycloguanil-d6, MedChemExpress
Description
Cycloguanil D6 is the deuterium labeled Cycloguanil, which is a dihydrofolate reductase inhibitor.
Specifications
Specifications
| Chemical Name or Material | Cycloguanil-d6 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C11H8D6ClN5 |
| Color | White |
| Synonym | Chlorguanide triazine-d6 |
| SMILES | ClC1=CC=C(N2C(N)=NC(N)=NC2(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1 |
| Molecular Weight (g/mol) | 257.75 |
| Quantity | 1 mg |
| Formula Weight | 257.75 |
| Show More |