Learn More
MedChemExpress Cyclo(-RGDfK)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Purity | 98.0% |
| CAS | 161552-03-0 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 603.67 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 125 mg/mL (207.07 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (82.83 mM∣Need ultrasonic) |