Learn More
Cyclin E1, p Thr77 Antibody (SR1828), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21555-50ul
 View more versions of this product
Product Code. 18663675
  • 338.00 EUR / 50µL
Description
Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclin E1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunofluorescence
SR1828
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
A synthesized peptide derived from human phospho-Cyclin E1 [Thr77] (Uniprot #: P24864)
50 μg
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific
898
Store at -20 degrees C. Avoid freeze/thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions