missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin C Polyclonal Antibody
GREENER_CHOICE

Product Code. 16391755
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16391755 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16391755 Supplier Invitrogen™ Supplier No. PA584810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GNFWQSSHYL QWILDKQDLL KERQKDLKFL SEEEYWKLQI FFTNVIQALG E Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Cyclin C was originally identified by a genetic screen for human and Drosophila cDNAs that complement a triple knock out of the CLN genes in Saccharomyces cerevisiae. Cyclin C is a nuclear protein present in a multiprotein complex. It interacts both in vitro and in vivo with cdk 8, a novel protein kinase of the cdk family. It is suggested that cyclin C and cdk 8 control RNA polymerase II function. Human cdk 8 cyclin C might be functionally associated with the mammalian transcription apparatus, perhaps involved in relaying growth regulatory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNC
Gene Accession No. P24863
Gene Alias AI451004; AU020987; CCNC; CG1C; CycC; cyclin C; cyclin-C; dJ199J3.4 (cyclin C); hSRB11; SRB11 homolog
Gene Symbols CCNC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCNC. Recombinant protein control fragment (Product #RP-105471).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.