missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1 Antibody (XLA1-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 EUR - 453.00 EUR
Specifications
| Antigen | Cyclin A1 |
|---|---|
| Clone | XLA1-1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18548260
|
Novus Biologicals
NB100-2624 |
0.1 mg |
453.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18411799
|
Novus Biologicals
NB100-2624-0.025mg |
0.025 mg |
190.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Xenopus samples. It is validated for Western BlotSpecifications
| Cyclin A1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS | |
| 8900 | |
| IgG1 | |
| Protein A purified |
| XLA1-1 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Xenopus species | |
| cyclin A1, cyclin-A1 | |
| Xenopus laevis cyclin A | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title