missing translation for 'onlineSavingsMsg'
Learn More

Cyanine 5, SE (Technical Grade), TRC

Product Code. 30359248
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30359248

Brand: Toronto Research Chemicals TRCC9605335MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanine 5, SE
CAS 146368-14-1
Molecular Formula C37 H43 N3 O10 S2
Quantity 5 mg
Synonym Cyanine 5, SE
InChI Formula InChI=1S/C37H43N3O10S2/c1-6-38-29-18-16-25(51(44,45)46)23-27(29)36(2,3)31(38)13-9-7-10-14-32-37(4,5)28-24-26(52(47,48)49)17-19-30(28)39(32)22-12-8-11-15-35(43)50-40-33(41)20-21-34(40)42/h7,9-10,13-14,16-19,23-24H,6,8,11-12,15,20-22H2,1-5H3,(H-,44,45,46,47,48,49)
SMILES CC[N+]1=C(\C=C\C=C\C=C/2\N(CCCCCC(=O)ON3C(=O)CCC3=O)c4ccc(cc4C2(C)C)S(=O)(=O)O)C(C)(C)c5cc(ccc15)S(=O)(=O)[O-]
IUPAC Name 2-[(1E,3E,5E)-5-[1-[6-(2,5-dioxopyrrolidin-1-yl)oxy-6-oxohexyl]-3,3-dimethyl-5-sulfoindol-2-ylidene]penta-1,3-dienyl]-1-ethyl-3,3-dimethylindol-1-ium-5-sulfonate
Molecular Weight (g/mol) 753.881
Formula Weight 753.239
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.