missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanine 5, SE (Technical Grade), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Cyanine 5, SE |
| CAS | 146368-14-1 |
| Molecular Formula | C37 H43 N3 O10 S2 |
| Quantity | 1 mg |
| Synonym | Cyanine 5, SE |
| InChI Formula | InChI=1S/C37H43N3O10S2/c1-6-38-29-18-16-25(51(44,45)46)23-27(29)36(2,3)31(38)13-9-7-10-14-32-37(4,5)28-24-26(52(47,48)49)17-19-30(28)39(32)22-12-8-11-15-35(43)50-40-33(41)20-21-34(40)42/h7,9-10,13-14,16-19,23-24H,6,8,11-12,15,20-22H2,1-5H3,(H-,44,45,46,47,48,49) |
| SMILES | CC[N+]1=C(\C=C\C=C\C=C/2\N(CCCCCC(=O)ON3C(=O)CCC3=O)c4ccc(cc4C2(C)C)S(=O)(=O)O)C(C)(C)c5cc(ccc15)S(=O)(=O)[O-] |
| IUPAC Name | 2-[(1E,3E,5E)-5-[1-[6-(2,5-dioxopyrrolidin-1-yl)oxy-6-oxohexyl]-3,3-dimethyl-5-sulfoindol-2-ylidene]penta-1,3-dienyl]-1-ethyl-3,3-dimethylindol-1-ium-5-sulfonate |
| Molecular Weight (g/mol) | 753.881 |
| Formula Weight | 753.239 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction