missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46112
This item is not returnable.
View return policy
Description
CXXC4 Polyclonal specifically detects CXXC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| CXXC4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CXXC finger protein 4, CXXC-type zinc finger protein 4, Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex), IDAXCXXC finger 4, Inhibition of the Dvl and axin complex protein, MGC149872 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 80319 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_079488.1 | |
| CXXC4 | |
| Peptide with sequence C-GTSLERTPVPSAEA corresponding to C-Terminus (near) according to NP_079488.1. | |
| 0.1 mg | |
| Primary | |
| Human, Expected from sequence similarity: Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction