missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7/RDC-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
226.00 EUR - 548.00 EUR
Specifications
| Antigen | CXCR7/RDC-1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 4 - 6 ug/mL, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Flow Cytometry (Negative) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18721664
|
Novus Biologicals
NBP2-24779SS |
0.025 mg |
226.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776883
|
Novus Biologicals
NBP2-24779 |
0.1 mg |
548.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR7/RDC-1 Polyclonal specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow Cytometry (Negative).Specifications
| CXCR7/RDC-1 | |
| Western Blot 4 - 6 ug/mL, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Flow Cytometry (Negative) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Sheep | |
| P25106 | |
| 57007 | |
| A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, GPCR | |
| PBS with 0.05% Sodium Azide | |
| chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| ACKR3 | |
| IgG | |
| Peptide affinity purified | |
| This antibody is reactive to Chimpanzee, Cow, Horse, Human, Monkey, Mouse, Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title