Learn More
Invitrogen™ CXCR7 Polyclonal Antibody

Description
PA3-069 detects CXC Chemokine Receptor Type 7 from human and mouse samples. PA3-069 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects a 42 kDa protein corresponding to mouse CXC Chemokine Receptor Type 7. The PA3-069 antigen is the COOH-terminal amino acids of mouse CXCR7 (347-362) SRVSETEYSALEQNTK coupled to KLH.
Specifications
Specifications
| Antigen | CXCR7 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.02% sodium azide |
| Gene | Ackr3 |
| Gene Accession No. | O89039, P25106, P56485 |
| Gene Alias | ACKR3; Atypical chemokine receptor 3; AW541270; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine; chemokine (C-X-C motif) receptor 7; chemokine orphan receptor 1; Cmkor1; CXC; C-X-C chemokine receptor type 7; CXC motif chemokine; CXCR7; CXC-R7; CXCR-7; G protein-coupled receptor; GPR159; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC1; RDC-1 |
| Gene Symbols | Ackr3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.