missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Rat anti-Mouse, PE, Clone: 12-81, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 566740
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCL16 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CXCL16 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 12-81 | |
| PE | |
| 0.1 mg | |
| Primary | |
| Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction