missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Rat anti-Mouse, Clone: 142417, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB503-SP
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL16 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58191 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 142417 | |
| Western Blot 1 ug/mL, Neutralization 0.15-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q8BSU2 | |
| CXCL16 | |
| E. coli-derived recombinant mouse CXCL16 Asn27-Pro114 Accession # Q8BSU2 | |
| 25 μg | |
| Primary | |
| Detects mouse CXCL16 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, recombinant mouse CXCL1, 2, 6, 9, 10, 11, 12, 13, 14, 15, recombinant rat CXCL1, 2, 3, 5, or recombinant porcine CXCL8 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction