missing translation for 'onlineSavingsMsg'
Learn More

CWF19L2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16843375 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16843375 20 μL 20µL
16833375 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16843375 Supplier Proteintech Supplier No. 248721AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CWF19L2 is a protein coding gene.

Specifications

Antigen CWF19L2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CWF19L2
Gene Accession No. Q2TBE0
Gene Alias CWF19 like protein 2, CWF19L2
Gene Symbols CWF19L2
Host Species Rabbit
Immunogen CWF19L2 Fusion Protein Ag21738
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143884
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.