missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eBioscience™

Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eBioscience™
Brand: Affymetrix eBioscience 14-9857-82
This item is not returnable.
View return policy
Description
Specifications
| Cutaneous Lymphocyte Antigen (CLA) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Rat | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot | |
| HECA-452 | |
| Unconjugated | |
| CLA | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction