Learn More
Ambion™ Custom Silencer™ siRNA
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Specifications
| Content And Storage | Room temperature |
| Description | Custom siRNA, 40nmol, HPLC Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA |
| Product Type | Custom siRNA |
| Purity | HPLC |
| Quantity | 40 nmol |