Learn More
Invitrogen™ Custom Silencer™ siRNA
Product Code. 15531144
Change view
Click to view available options
Description:
Custom siRNA HPLC, 250 nmol
Custom siRNA, 10 umol
Custom siRNA, 20 nmol
Custom siRNA, 250 nmol
Unit Size:
10µmol
20nmol
250nmol
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15531144 Custom siRNA, 20 nmol 20nmol
15571064 Custom siRNA HPLC, 250 nmol 250nmol
15511154 Custom siRNA, 250 nmol 250nmol
15531154 Custom siRNA, 10 umol 10µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15531144 Supplier Invitrogen™ Supplier No. AM16100
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types.
Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -20°C. Do not store in a frost-free freezer. Dried oligonucleotides are shipped at ambient temperature.
Description Custom siRNA, 20 nmol
RNAi Type siRNA
Sample Type RNA
Product Line Ambion™, Silencer™ Select
Product Type siRNA
Quantity 20 nmol
For Research Use Only. Not for use in diagnostic procedures.