missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ Cultrex Poly-D-Lysine
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Substrate for cell culture adhesion. An area of 25 cm2 can be coated with 0.5 mL of a 0.01% Poly-D-Lysine solution. Optimal conditions for attachment must be determined for each cell line and application. Slides may be dipped in the solution and air dried before applying sample.
Specifications
Specifications
| Content And Storage | Product is stable for at least 6 months from the date of receipt when stored at 2 - 8 °C. Keep sterile. Sterility Testing: No bacterial or fungal growth detected after incubation at 37 °C for 14 days following USP Chapter 71 sterility testing. |
| For Use With (Application) | Bioactivity |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction