Learn More
Thermo Scientific™ Culti-Loops™ Yersinia enterocolitica subsp. enterocolitica ATCC™ 9610™
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 9610TM* |
| Description | Yersinia enterocolitica subsp. enterocolitica ATCC™ 9610™ |
| Format | Culti-Loops |
| Product Type | Culti-Loops Quality Control Organism |
| Target Organism Class | Gram-negative Rods |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Organism |
| Strain Designations | 33114 [CCUG 11291, CCUG 12369, CIP 80.27, DSM 4780, LMG 7899, NCTC 12982] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.