missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Culti-Loops™ Haemophilus influenzae ATCC™ 33930™

Product Code. 12969008
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12969008 5 Loops/Pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12969008 Supplier Thermo Scientific™ Supplier No. R4603903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 33930TM*
Description Haemophilus influenzae ATCC™ 33930™; Beta-lactamase positive
Format Culti-Loops
Product Type Culti-Loop
Target Organism Class Fastidious
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations R387

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.